Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046193.1 | XP_017901682.1 | 2 - 3 | 1099 | 88 | |
GenBank mRNA | JT669282.1 | - | 3 - 4 | 945 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |