Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674693.2 | XP_005674750.2 | 3 - 4 | 540 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |