Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676315.3 | XP_005676372.2 | 3 - 4 | 3414 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |