Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677436.3 | XP_005677493.1 | 6 - 7 | 1714 | 89 | |
XM_005677437.3 | XP_005677494.1 | 6 - 7 | 949 | 89 | ||
GenBank mRNA | JO440784.2 | - | 6 - 7 | 816 | 89 | |
KA344686.1 | - | 5 - 6 | 839 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |