Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679826.3 | XP_005679883.1 | 1 - 2 | 144 | 79 | |
XM_005679827.3 | XP_005679884.1 | 1 - 2 | 134 | 79 | ||
GenBank mRNA | JO453432.2 | - | 3 - 4 | 454 | 79 | |
JO611279.2 | - | 1 - 2 | 111 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |