Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687268.3 | XP_005687325.2 | 3 - 4 | 737 | 75 | |
XM_018055958.1 | XP_017911447.1 | 3 - 4 | 719 | 75 | ||
XM_018055959.1 | XP_017911448.1 | 3 - 4 | 719 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |