Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689373.3 | XP_005689430.2 | 6 - 7 | 702 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |