Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061515.1 | XP_017917004.1 | 2 - 3 | 1284 | 64 | |
GenBank mRNA | JT709803.1 | - | 3 - 4 | 71 |
Set Membership: |
Amplicon length less than or equal to 70 Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |