Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042811.1 | XP_017898300.1 | 4 - 5 | 739 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |