Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013967282.2 | XP_013822736.2 | 10 - 11 | 3178 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |