Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013967282.2 | XP_013822736.2 | 9 - 10 | 2989 | 107 | |
XM_018053995.1 | XP_017909484.1 | 9 - 10 | 2989 | 107 | ||
GenBank mRNA | JO454271.2 | - | 3 - 4 | 388 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |