Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676525.3 | XP_005676582.2 | 8 - 9 | 1773 | 62 | |
GenBank mRNA | JO321018.2 | - | 1 - 2 | 529 | 62 | |
JR128856.1 | - | 4483 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |