Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676604.3 | XP_005676661.1 | 5 - 6 | 1174 | 58 | |
GenBank mRNA | JQ407589.1 | - | 1 - 2 | 178 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |