Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680207.2 | XP_005680264.2 | 2 - 3 | 339 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |