Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14H8orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689042.3 | XP_005689099.2 | 9 - 10 | 1538 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |