Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692107.3 | XP_005692164.1 | 8 - 9 | 889 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |