Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048636.1 | XP_017904125.1 | 5 - 6 | 910 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |