Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973756.2 | XP_013829210.2 | 2 - 3 | 307 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |