Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680013.2 | XP_005680070.1 | 2 - 3 | 313 | 69 | |
GenBank mRNA | JO453992.2 | - | 2 - 3 | 316 | 69 | |
JO623831.2 | - | 3 - 4 | 1318 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |