Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047680.1 | XP_017903169.1 | 2 - 3 | 378 | 79 | |
GenBank mRNA | JO639605.2 | - | 5 - 6 | 889 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |