Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048413.1 | XP_017903902.1 | 6 - 7 | 1431 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |