Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686088.3 | XP_005686145.1 | 1 - 2 | 574 | 104 | |
XM_005686089.3 | XP_005686146.1 | 1 - 2 | 563 | 104 | ||
GenBank mRNA | KA338182.1 | - | 1 - 2 | 394 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |