Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686088.3 | XP_005686145.1 | 6 - 7 | 980 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |