Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686638.3 | XP_005686695.2 | 1 - 2 | 139 | 80 | |
GenBank mRNA | JT680139.1 | - | 7 - 8 | 1319 | 80 | |
JT701139.1 | - | 9 - 10 | 1364 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |