Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692041.1 | XP_005692098.1 | 4 - 5 | 665 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |