Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MILR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694010.3 | XP_005694067.2 | 2 - 3 | 541 | 95 | |
XM_018065359.1 | XP_017920848.1 | 2 - 3 | 541 | 95 | ||
GenBank mRNA | JO595577.2 | - | 2 - 3 | 451 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |