Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698843.3 | XP_005698900.1 | 2 - 3 | 537 | 107 | |
GenBank mRNA | JT753549.1 | - | 5 - 6 | 1011 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |