Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699144.3 | XP_005699201.2 | 4 - 5 | 577 | 98 | |
XM_018042587.1 | XP_017898076.1 | 5 - 6 | 588 | 98 | ||
GenBank mRNA | JI861768.1 | - | 3 - 4 | 396 | 98 | |
JO449895.2 | - | 4 - 5 | 567 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |