Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051240.1 | XP_017906729.1 | 5 - 6 | 1526 | 81 | |
GenBank mRNA | JO577993.1 | - | 1 - 2 | 1215 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |