Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013967834.2 | XP_013823288.1 | 2 - 3 | 239 | 65 | |
XM_018055581.1 | XP_017911070.1 | 2 - 3 | 439 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |