Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058448.1 | XP_017913937.1 | 4 - 5 | 762 | 74 | |
GenBank mRNA | JO636843.2 | - | 4 - 5 | 577 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |