Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066825.1 | XP_017922314.1 | 4 - 5 | 1972 | 93 | |
XM_018066826.1 | XP_017922315.1 | 4 - 5 | 1851 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |