Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066628.1 | XP_017922117.1 | 3 - 4 | 2046 | 88 | |
GenBank mRNA | KA334310.1 | - | 2 - 3 | 966 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |