Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066628.1 | XP_017922117.1 | 4 - 5 | 2212 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |