Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679653.3 | XP_005679710.1 | 2 - 3 | 1685 | 64 | |
GenBank mRNA | JO420810.2 | - | 2 - 3 | 1674 | 64 | |
JO613184.2 | - | 3 - 4 | 1459 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |