Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049093.1 | XP_017904582.1 | 2 - 3 | 814 | 102 | |
GenBank mRNA | JT652264.1 | - | 3 - 4 | 376 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |