Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680979.3 | XP_005681036.2 | 3 - 4 | 836 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |