Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683598.3 | XP_005683655.1 | 6 - 7 | 812 | 108 | |
XM_005683599.2 | XP_005683656.1 | 5 - 6 | 563 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |