Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLANA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683687.2 | XP_005683744.1 | 4 - 5 | 364 | 81 | |
GenBank mRNA | KA320474.1 | - | 4 - 5 | 378 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |