Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691946.3 | XP_005692003.1 | 3 - 4 | 280 | 85 | |
GenBank mRNA | JO426407.1 | - | 14 - 15 | 2895 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |