Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064531.1 | XP_017920020.1 | 4 - 5 | 452 | 85 | |
GenBank mRNA | JO611770.2 | - | 1 - 2 | 425 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |