Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966979.2 | XP_013822433.2 | 1 - 2 | 656 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |