Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060653.1 | XP_017916142.1 | 2 - 3 | 463 | 78 | |
GenBank mRNA | JO354498.2 | - | 2 - 3 | 179 | 78 | |
JO433014.2 | - | 2 - 3 | 469 | 78 | ||
JO587247.2 | - | 2 - 3 | 508 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |