Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971456.2 | XP_013826910.2 | 3 - 4 | 1943 | 56 | |
GenBank mRNA | KA329329.1 | - | 1 - 2 | 148 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |