Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066260.1 | XP_017921749.1 | 3 - 4 | 1756 | 79 | |
XM_018066261.1 | XP_017921750.1 | 2 - 3 | 1573 | 79 | ||
XM_018066266.1 | XP_017921755.1 | 4 - 5 | 1864 | 79 | ||
GenBank mRNA | JO646847.2 | - | 1 - 2 | 121 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |