Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO405679.2 | - | 3 - 4 | 935 | 76 | |
JT762829.1 | - | 1 - 2 | 49 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |