Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046241.1 | XP_017901730.1 | 1 - 2 | 621 | 54 | |
GenBank mRNA | JO633353.2 | - | 1 - 2 | 298 | 54 | |
KA324385.1 | - | 8 - 9 | 1283 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |