Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043628.1 | XP_017899117.1 | 5 - 6 | 1353 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |