Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054573.1 | XP_017910062.1 | 1 - 2 | 1669 | 54 | |
XM_018054574.1 | XP_017910063.1 | 2 - 3 | 3639 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |