Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686599.3 | XP_005686656.1 | 2 - 3 | 465 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |